siRNA Details
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122951.2 | NP_001116423.1 | 1 | 256 | ||
| NM_002036.3 | NP_002027.2 | 2 | 1022 | |||
| GenBank mRNA | AF030521.1 | - | 1 | 81 | ||
| AK291593.1 | - | 2 | 1022 | |||
| AK309521.1 | - | 1 | 525 | |||
| AY167991.1 | - | 1 | 75 | |||
| BC017817.1 | - | 2 | 227 | 4696425 | ||
| BC095496.1 | - | 1 | 78 | |||
| BX537430.1 | - | 3 | 919 | |||
| CD701182.1 | - | 2 | 336 | |||
| EL735542.1 | - | 2 | 75 | |||
| HE572751.1 | - | 2 | 75 | |||
| JX081310.1 | - | 1 | 81 | |||
| U01839.1 | - | 1 | 256 | |||

ACKR1
Genomic Map