siRNA Details
| Gene Symbol: FAM90A10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046351.1 | 3 | 384 | |||
| GenBank mRNA | DC395395.1 | - | 3 | 385 | ||
| Gene Symbol: FAM90A7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046343.1 | 3 | 384 | |||
| GenBank mRNA | AK301810.1 | - | 3 | 385 | ||

FAM90A10P
Genomic Map