siRNA Details
| Gene Symbol: AARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025357.2 | NP_001020528.1 | 2 | 408 | ||
| XM_011517043.2 | XP_011515345.1 | 2 | 786 | |||
| GenBank mRNA | AB181519.1 | - | 2 | 389 | ||
| BC140924.1 | - | 2 | 434 | 9056736 | ||
| BC142621.1 | - | 2 | 408 | 40147448 | ||

AARD
Genomic Map