siRNA Details
| Gene Symbol: NUTM2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045477.2 | NP_001038942.1 | 3 | 843 | ||
| NM_001170741.1 | NP_001164212.1 | 3 | 843 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUTM2G| Gene Symbol: NUTM2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045477.2 | NP_001038942.1 | 3 | 843 | ||
| NM_001170741.1 | NP_001164212.1 | 3 | 843 | |||