siRNA Details
| Gene Symbol: NRARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 1338 | ||
| GenBank mRNA | BC053618.1 | - | 1 | 1338 | 6143133 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NRARP| Gene Symbol: NRARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 1338 | ||
| GenBank mRNA | BC053618.1 | - | 1 | 1338 | 6143133 | |