siRNA Details
| Gene Symbol: CENPVP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033772.1 | 1 | 1022 | |||
| GenBank mRNA | AY730278.1 | - | 1 | 1022 | ||
| Gene Symbol: CENPVP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033773.1 | 1 | 1022 | |||

CENPVP1
Genomic Map