siRNA Details
| Gene Symbol: MFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033540.2 | NP_284941.2 | 16 | 1981 | ||
| XM_005247596.3 | XP_005247653.2 | 16 | 1939 | |||
| XM_011512963.2 | XP_011511265.1 | 13 | 1567 | |||
| GenBank mRNA | AB043588.1 | - | 15 | 1946 | ||
| AB043589.1 | - | 16 | 1946 | |||
| AF054986.1 | - | 6 | 738 | |||
| AF329637.1 | - | 15 | 1855 | |||
| AK000700.1 | - | 15 | 1811 | |||
| AK314306.1 | - | 16 | 1938 | |||
| BC040557.1 | - | 16 | 1981 | 5270347 | ||
| CR627379.1 | - | 13 | 1630 | |||
| U95822.1 | - | 10 | 1111 | |||

MFN1
Genomic Map