siRNA Details
Gene Symbol: FAM127C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078173.1 | NP_001071641.1 | 1 | 1302 | ||
GenBank mRNA | BC048268.1 | - | 1 | 945 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM127C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078173.1 | NP_001071641.1 | 1 | 1302 | ||
GenBank mRNA | BC048268.1 | - | 1 | 945 |