siRNA Details
| Gene Symbol: CT45A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017438.2 | NP_001017438.2 | 5 | 774 | ||
| XM_005262425.4 | XP_005262482.2 | 5 | 694 | |||
| XM_011531351.1 | XP_011529653.1 | 5 | 625 | |||
| GenBank mRNA | AY743714.1 | - | 5 | 774 | ||
| Gene Symbol: CT45A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017417.2 | NP_001017417.1 | 5 | 773 | ||
| XM_005278141.3 | XP_005278198.1 | 5 | 622 | |||
| XM_011531352.1 | XP_011529654.1 | 4 | 543 | |||
| GenBank mRNA | AY743709.1 | - | 5 | 774 | ||
| BC140913.1 | - | 5 | 673 | 9056725 | ||
| Gene Symbol: CT45A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291527.1 | NP_001278456.1 | 5 | 698 | ||
| NM_001291528.1 | NP_001278457.1 | 5 | 681 | |||
| NM_001291529.1 | NP_001278458.1 | 5 | 619 | |||
| NM_001291530.1 | NP_001278459.1 | 5 | 636 | |||
| XM_011531238.2 | XP_011529540.1 | 5 | 632 | |||
| XM_011531239.1 | XP_011529541.1 | 5 | 613 | |||
| GenBank mRNA | AK292207.1 | - | 5 | 615 | ||
| AY743713.1 | - | 5 | 774 | |||
| BC070138.1 | - | 5 | 619 | 4685945 | ||
| BC144429.1 | - | 5 | 673 | 9052950 | ||
| BC144430.1 | - | 5 | 690 | 9052951 | ||
| BC144527.1 | - | 5 | 696 | 9053054 | ||
| BC144528.1 | - | 5 | 681 | 9053055 | ||
| Gene Symbol: CT45A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152582.6 | NP_689795.4 | 5 | 774 | ||
| XM_006724783.2 | XP_006724846.1 | 5 | 701 | |||
| XM_011531389.2 | XP_011529691.1 | 5 | 669 | |||
| GenBank mRNA | AY743710.1 | - | 5 | 774 | ||
| Gene Symbol: CT45A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017435.1 | NP_001017435.1 | 5 | 774 | ||
| XM_006724758.3 | XP_006724821.1 | 5 | 691 | |||
| XM_011531340.2 | XP_011529642.1 | 5 | 650 | |||
| GenBank mRNA | AK098689.1 | - | 5 | 619 | ||
| AY743711.1 | - | 5 | 774 | |||
| Gene Symbol: CT45A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007551.4 | NP_001007552.2 | 5 | 774 | ||
| NM_001172288.2 | NP_001165759.2 | 5 | 791 | |||
| XM_011531341.2 | XP_011529643.1 | 5 | 656 | |||
| GenBank mRNA | BC028711.2 | - | 5 | 774 | 4828274 | |
| BC140735.1 | - | 5 | 681 | 9021603 | ||
| BC140926.1 | - | 5 | 681 | 9056738 | ||
| BC144437.1 | - | 5 | 681 | 9052959 | ||
| Gene Symbol: CT45A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291543.1 | NP_001278472.1 | 5 | 590 | ||
| XM_003960082.4 | XP_003960131.1 | 5 | 677 | |||
| XM_011531229.2 | XP_011529531.1 | 5 | 656 | |||
| GenBank mRNA | BU540102.1 | - | 5 | 593 | ||
| Gene Symbol: CT45A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291535.1 | NP_001278464.1 | 5 | 774 | ||
| XM_006724799.2 | XP_006724862.1 | 5 | 701 | |||
| XM_011531241.2 | XP_011529543.1 | 5 | 669 | |||
| Gene Symbol: CT45A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291540.2 | NP_001278469.1 | 5 | 774 | ||
| NM_001321271.1 | NP_001308200.1 | 5 | 619 | |||
| XM_006724796.2 | XP_006724859.1 | 5 | 701 | |||

CT45A6
Genomic Map