siRNA Details
| Gene Symbol: FRG2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080998.1 | NP_001074467.1 | 4 | 410 | ||
| GenBank mRNA | AY744466.1 | - | 4 | 410 | ||
| Gene Symbol: FRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005217.1 | NP_001005217.1 | 4 | 410 | ||
| NM_001286820.1 | NP_001273749.1 | 4 | 413 | |||
| GenBank mRNA | AY714545.1 | - | 4 | 410 | ||
| BC144572.1 | - | 4 | 398 | 9053101 | ||

FRG2B
Genomic Map