siRNA Details
| Gene Symbol: TSPAN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080509.2 | NP_001073978.1 | 2 | 157 | ||
| XM_011520679.2 | XP_011518981.1 | 2 | 419 | |||
| XM_011520680.2 | XP_011518982.1 | 2 | 397 | |||
| GenBank mRNA | AK293947.1 | - | 2 | 104 | ||
| BC084571.1 | - | 2 | 424 | |||
| BC146917.1 | - | 2 | 103 | 9056845 | ||
| BC146922.1 | - | 2 | 103 | 9056850 | ||

TSPAN11
Genomic Map