siRNA Details
| Gene Symbol: TSPAN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080509.2 | NP_001073978.1 | 6 | 624 | ||
| XM_011520679.2 | XP_011518981.1 | 6 | 886 | |||
| XM_011520680.2 | XP_011518982.1 | 6 | 864 | |||
| GenBank mRNA | AK293947.1 | - | 6 | 571 | ||
| AK300920.1 | - | 5 | 529 | |||
| BC127660.1 | - | 4 | 390 | |||
| BC127661.1 | - | 4 | 390 | |||
| BC146917.1 | - | 6 | 570 | 9056845 | ||
| BC146922.1 | - | 6 | 570 | 9056850 | ||

TSPAN11
Genomic Map