siRNA Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002038.3 | NP_002029.3 | 5 | 759 | ||
| NM_022872.2 | NP_075010.1 | 5 | 771 | |||
| NM_022873.2 | NP_075011.1 | 5 | 783 | |||
| GenBank mRNA | AK024814.1 | - | 5 | 738 | ||
| BC011601.2 | - | 5 | 711 | 3140035 | ||
| BC015603.2 | - | 5 | 714 | 4654295 | ||
| X02492.1 | - | 5 | 705 | |||

IFI6
Genomic Map