siRNA Details
| Gene Symbol: BECN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290693.1 | NP_001277622.1 | 1 | 1189 | ||
| GenBank mRNA | HM031116.1 | - | 1 | 1189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BECN2| Gene Symbol: BECN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290693.1 | NP_001277622.1 | 1 | 1189 | ||
| GenBank mRNA | HM031116.1 | - | 1 | 1189 | ||