siRNA Details
| Gene Symbol: OR13G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005487.1 | NP_001005487.1 | 1 | 759 | ||
| GenBank mRNA | BC136736.1 | - | 1 | 781 | 9020726 | |
| BC136738.1 | - | 1 | 781 | 9020728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR13G1| Gene Symbol: OR13G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005487.1 | NP_001005487.1 | 1 | 759 | ||
| GenBank mRNA | BC136736.1 | - | 1 | 781 | 9020726 | |
| BC136738.1 | - | 1 | 781 | 9020728 | ||