siRNA Details
| Gene Symbol: SULT1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008743.2 | NP_001008743.1 | 2 | 187 | ||
| NM_001320878.1 | NP_001307807.1 | 2 | 187 | |||
| XM_017004153.1 | XP_016859642.1 | 1 | 1076 | |||
| XM_017004154.1 | XP_016859643.1 | 1 | 1076 | |||
| XM_017004155.1 | XP_016859644.1 | 1 | 1076 | |||

SULT1C3
Genomic Map