siRNA Details
Gene Symbol: OR10C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013941.3 | NP_039229.3 | 1 | 738 | ||
GenBank mRNA | BC136996.1 | - | 1 | 738 | 9020988 | |
BC136997.1 | - | 1 | 738 | 9020989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR10C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013941.3 | NP_039229.3 | 1 | 738 | ||
GenBank mRNA | BC136996.1 | - | 1 | 738 | 9020988 | |
BC136997.1 | - | 1 | 738 | 9020989 |