siRNA Details
| Gene Symbol: SLC37A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164277.1 | NP_001157749.1 | 6, 7 | 1541 | ||
| NM_001164278.1 | NP_001157750.1 | 6, 7 | 1225 | |||
| NM_001164279.1 | NP_001157751.1 | 6, 7 | 967 | |||
| NM_001164280.1 | NP_001157752.1 | 4, 5 | 983 | |||
| NM_001467.5 | NP_001458.1 | 5, 6 | 1037 | |||
| GenBank mRNA | AF110819.1 | - | 4, 5 | 784 | ||
| AF110820.1 | - | 4, 5 | 784 | |||
| AF110822.1 | - | 4, 5 | 784 | |||
| AF111852.1 | - | 2, 3 | 1537 | |||
| AK289642.1 | - | 6, 7 | 1509 | |||
| AK298395.1 | - | 6, 7 | 921 | |||
| AK300646.1 | - | 6, 7 | 933 | |||
| AK312628.1 | - | 6, 7 | 1207 | |||
| AY423732.1 | - | 4, 5 | 804 | |||
| BC002400.2 | - | 4, 5 | 985 | 2822215 | ||
| BC003589.1 | - | 4, 5 | 985 | 2822215 | ||
| BC014663.1 | - | 4, 5 | 992 | 4857067 | ||
| BC015650.2 | - | 5, 6 | 1457 | 4660508 | ||
| BC064563.1 | - | 4, 5 | 980 | 5580134 | ||
| CR456733.1 | - | 4, 5 | 784 | |||
| Y15409.1 | - | 4, 5 | 953 | |||

SLC37A4
Genomic Map