siRNA Details
| Gene Symbol: FAM47C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013736.2 | NP_001013758.1 | 1 | 2774 | ||
| GenBank mRNA | AK125992.1 | - | 1 | 2496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM47C| Gene Symbol: FAM47C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013736.2 | NP_001013758.1 | 1 | 2774 | ||
| GenBank mRNA | AK125992.1 | - | 1 | 2496 | ||