siRNA Details
| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040663.2 | NP_001035753.1 | 5 | 711 | ||
| NR_102272.1 | 6 | 854 | ||||
| GenBank mRNA | BC036094.1 | - | 6 | 842 | 5296612 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAGE1| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040663.2 | NP_001035753.1 | 5 | 711 | ||
| NR_102272.1 | 6 | 854 | ||||
| GenBank mRNA | BC036094.1 | - | 6 | 842 | 5296612 | |