siRNA Details
| Gene Symbol: BPY2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002761.1 | NP_001002761.1 | 5 | 459 | ||
| Gene Symbol: BPY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004678.2 | NP_004669.2 | 5 | 459 | ||
| GenBank mRNA | AF000980.1 | - | 5 | 459 | ||
| Gene Symbol: BPY2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002760.1 | NP_001002760.1 | 5 | 459 | ||

BPY2C
Genomic Map