siRNA Details
| Gene Symbol: LRRC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006939.3 | NP_001006940.3 | 1 | 254 | ||
| XM_006717867.2 | XP_006717930.1 | 3 | 731 | |||
| XM_011539822.1 | XP_011538124.1 | 4 | 867 | |||
| XM_011539823.1 | XP_011538125.1 | 3 | 734 | |||
| XM_011539826.2 | XP_011538128.1 | 2 | 2021 | |||
| XM_011539828.2 | XP_011538130.1 | 2 | 1560 | |||
| XM_011539829.2 | XP_011538131.1 | 2 | 1563 | |||
| XM_017016276.1 | XP_016871765.1 | 4 | 870 | |||
| XM_017016277.1 | XP_016871766.1 | 2 | 2024 | |||
| GenBank mRNA | AY358137.1 | - | 2 | 275 | ||
| BC036722.1 | - | 1 | 265 | 5206652 | ||

LRRC18
Genomic Map