siRNA Details
| Gene Symbol: LRRC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006939.3 | NP_001006940.3 | 1 | 333 | ||
| XM_006717867.2 | XP_006717930.1 | 3 | 810 | |||
| XM_011539822.1 | XP_011538124.1 | 4 | 946 | |||
| XM_011539823.1 | XP_011538125.1 | 3 | 813 | |||
| XM_011539826.2 | XP_011538128.1 | 2 | 2100 | |||
| XM_011539828.2 | XP_011538130.1 | 2 | 1639 | |||
| XM_011539829.2 | XP_011538131.1 | 2 | 1642 | |||
| XM_017016276.1 | XP_016871765.1 | 4 | 949 | |||
| XM_017016277.1 | XP_016871766.1 | 2 | 2103 | |||
| GenBank mRNA | AY358137.1 | - | 2 | 354 | ||
| BC036722.1 | - | 1 | 344 | 5206652 | ||

LRRC18
Genomic Map