siRNA Details
| Gene Symbol: TRIM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090695.1 | - | 2 | 164 | ||
| AK131485.1 | - | 3 | 664 | |||
| BC033211.1 | - | 3 | 649 | 4906353 | ||
| DA058421.1 | - | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM72| Gene Symbol: TRIM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090695.1 | - | 2 | 164 | ||
| AK131485.1 | - | 3 | 664 | |||
| BC033211.1 | - | 3 | 649 | 4906353 | ||
| DA058421.1 | - | 2 | 164 | |||