siRNA Details
| Gene Symbol: PHOSPHO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008489.3 | NP_001008489.1 | 4 | 641 | ||
| NM_001199285.1 | NP_001186214.1 | 4 | 541 | |||
| NM_001199286.1 | NP_001186215.1 | 4 | 760 | |||
| NM_001199287.1 | NP_001186216.1 | 5 | 759 | |||
| NM_001199288.1 | NP_001186217.1 | 4 | 560 | |||
| GenBank mRNA | AK314915.1 | - | 4 | 544 | ||
| BC022324.2 | - | 3 | 472 | 4701753 | ||
| BC106013.1 | - | 4 | 630 | 4387304 | ||

PHOSPHO2
Genomic Map