siRNA Details
| Gene Symbol: SPANXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009615.2 | NP_001009615.1 | 2 | 139 | ||
| GenBank mRNA | DQ336111.1 | - | 2 | 139 | ||
| DQ336112.1 | - | 2 | 139 | |||
| DQ336113.1 | - | 2 | 139 | |||
| DQ336114.1 | - | 2 | 139 | |||
| DQ336115.1 | - | 2 | 139 | |||

SPANXN2
Genomic Map