siRNA Details
| Gene Symbol: C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035005.3 | NP_001030177.1 | 2 | 390 | ||
| NM_001199346.1 | NP_001186275.1 | 2 | 226 | |||
| GenBank mRNA | AB097012.1 | - | 2 | 310 | ||
| AK027111.1 | - | 2 | 186 | |||
| AK291882.1 | - | 2 | 197 | |||
| BC022357.1 | - | 2 | 195 | 4297017 | ||
| BC093004.1 | - | 2 | 347 | 5273608 | ||
| BF210402.1 | - | 2 | 225 | |||
| Gene Symbol: RPL17-C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199355.1 | NP_001186284.1 | 6 | 678 | ||
| NM_001199356.1 | NP_001186285.1 | 6 | 745 | |||
| GenBank mRNA | BX393840.2 | - | 6 | 680 | ||

C18orf32
Genomic Map