siRNA Details
| Gene Symbol: C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035005.3 | NP_001030177.1 | 3 | 548 | ||
| NM_001199346.1 | NP_001186275.1 | 3 | 384 | |||
| GenBank mRNA | AB097012.1 | - | 3 | 468 | ||
| AK027111.1 | - | 3 | 344 | |||
| AK291882.1 | - | 3 | 355 | |||
| BC022357.1 | - | 3 | 353 | 4297017 | ||
| BC093004.1 | - | 3 | 505 | 5273608 | ||
| BF210402.1 | - | 3 | 383 | |||
| Gene Symbol: RPL17-C18orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199355.1 | NP_001186284.1 | 7 | 836 | ||
| NM_001199356.1 | NP_001186285.1 | 7 | 903 | |||
| GenBank mRNA | BX393840.2 | - | 7 | 838 | ||

C18orf32
Genomic Map