siRNA Details
| Gene Symbol: DEFB107B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040705.1 | NP_001035795.1 | 2 | 186 | ||
| Gene Symbol: DEFB107A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037668.1 | NP_001032757.2 | 2 | 186 | ||
| GenBank mRNA | AF540979.1 | - | 2 | 86 | ||
| AY122467.1 | - | 2 | 86 | |||
| BC137510.1 | - | 2 | 112 | 9021510 | ||
| BC137513.1 | - | 2 | 112 | 9021513 | ||
| DQ119826.1 | - | 2 | 186 | |||

DEFB107B
Genomic Map