siRNA Details
| Gene Symbol: DEFB104B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040702.1 | NP_001035792.1 | 2 | 184 | ||
| Gene Symbol: DEFB104A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080389.2 | NP_525128.2 | 2 | 184 | ||
| GenBank mRNA | AJ314834.1 | - | 2 | 184 | ||
| BC100848.1 | - | 2 | 184 | 40002160 | ||
| BC100849.1 | - | 2 | 184 | 40002161 | ||
| BC100850.1 | - | 2 | 184 | 40002162 | ||
| BC100851.1 | - | 2 | 184 | 40002163 | ||

DEFB104B
Genomic Map