siRNA Details
| Gene Symbol: YIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039671.2 | NP_001034760.1 | 8 | 2662 | ||
| NM_001039672.2 | NP_001034761.1 | 8 | 2680 | |||
| NM_001039673.2 | NP_001034762.1 | 8 | 2671 | |||
| NM_001145461.1 | NP_001138933.1 | 8 | 2656 | |||
| NM_001145462.1 | NP_001138934.1 | 8 | 2632 | |||
| NM_001145463.1 | NP_001138935.1 | 9 | 2748 | |||
| GenBank mRNA | AK027130.1 | - | 1 | 1355 | ||
| BC025257.1 | - | 1 | 1291 | |||

YIF1B
Genomic Map