siRNA Details
| Gene Symbol: CLLU1OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025232.1 | NP_001020403.1 | 4 | 591 | ||
| XM_017019673.1 | XP_016875162.1 | 4 | 877 | |||
| GenBank mRNA | AJ845168.1 | - | 4 | 591 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLLU1OS| Gene Symbol: CLLU1OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025232.1 | NP_001020403.1 | 4 | 591 | ||
| XM_017019673.1 | XP_016875162.1 | 4 | 877 | |||
| GenBank mRNA | AJ845168.1 | - | 4 | 591 | ||