siRNA Details
| Gene Symbol: CLLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025233.2 | NP_001020404.1 | 1 | 844 | ||
| GenBank mRNA | AJ845165.1 | - | 1 | 844 | ||
| AJ845166.1 | - | 1 | 844 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLLU1| Gene Symbol: CLLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025233.2 | NP_001020404.1 | 1 | 844 | ||
| GenBank mRNA | AJ845165.1 | - | 1 | 844 | ||
| AJ845166.1 | - | 1 | 844 | |||