siRNA Details
| Gene Symbol: NPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030013.1 | NP_001025184.1 | 3 | 215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPS| Gene Symbol: NPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030013.1 | NP_001025184.1 | 3 | 215 | ||