siRNA Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031672.2 | NP_001026842.2 | 4 | 588 | ||
| GenBank mRNA | AK090456.1 | - | 4 | 526 | ||
| BC064419.1 | - | 4 | 551 | |||
| BC071735.1 | - | 4 | 527 | |||
| CD515193.1 | - | 2 | 334 | |||

CYB5RL
Genomic Map