siRNA Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031672.2 | NP_001026842.2 | 5 | 695 | ||
| GenBank mRNA | AK090456.1 | - | 5 | 786 | ||
| BC012781.2 | - | 4 | 479 | |||
| BC049198.1 | - | 2 | 203 | |||
| BC064419.1 | - | 5 | 668 | |||
| BC071735.1 | - | 5 | 644 | |||
| CD515193.1 | - | 3 | 441 | |||

CYB5RL
Genomic Map