siRNA Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031672.2 | NP_001026842.2 | 4, 5 | 678 | ||
| GenBank mRNA | AK090456.1 | - | 5 | 769 | ||
| BC012781.2 | - | 3, 4 | 462 | |||
| BC049198.1 | - | 1, 2 | 186 | |||
| BC064419.1 | - | 4, 5 | 651 | |||
| BC071735.1 | - | 4, 5 | 627 | |||
| CD515193.1 | - | 2, 3 | 424 | |||
| DA793739.1 | - | 3, 4 | 529 | |||

CYB5RL
Genomic Map