siRNA Details
| Gene Symbol: ARL17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113738.1 | NP_001107210.1 | 2 | 242 | ||
| NM_001288811.1 | NP_001275740.1 | 2 | 242 | |||
| NM_001288812.1 | NP_001275741.1 | 2 | 242 | |||
| NM_001288813.1 | NP_001275742.1 | 2 | 242 | |||
| NM_016632.2 | NP_057716.2 | 2 | 242 | |||
| XM_005257439.4 | XP_005257496.1 | 2 | 301 | |||
| XM_011524875.2 | XP_011523177.1 | 2 | 295 | |||
| XM_011524876.2 | XP_011523178.1 | 2 | 294 | |||
| XM_011524877.2 | XP_011523179.1 | 2 | 290 | |||
| XM_011524878.2 | XP_011523180.1 | 2 | 292 | |||
| XM_017024729.1 | XP_016880218.1 | 2 | 294 | |||
| GenBank mRNA | AB209344.1 | - | 2 | 173 | ||
| AF208864.1 | - | 2 | 190 | |||
| AF493886.1 | - | 1 | 103 | |||
| AK289590.1 | - | 2 | 180 | |||
| AK315857.1 | - | 2 | 181 | |||
| BC020869.1 | - | 2 | 163 | |||
| BC030570.1 | - | 2 | 213 | 5518676 | ||
| BC041803.1 | - | 2 | 199 | 5303659 | ||
| BX647266.1 | - | 2 | 202 | |||
| DA761795.1 | - | 2 | 192 | |||
| DA979569.1 | - | 2 | 174 | |||
| DB027627.1 | - | 2 | 203 | |||
| DB065209.1 | - | 2 | 242 | |||
| DB241677.1 | - | 2 | 222 | |||
| Gene Symbol: ARL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039083.3 | NP_001034172.3 | 2 | 213 | ||
| NM_001103154.1 | NP_001096624.1 | 2 | 213 | |||
| XM_005256918.4 | XP_005256975.1 | 2 | 317 | |||
| XM_011524160.2 | XP_011522462.1 | 2 | 313 | |||
| XM_011524161.2 | XP_011522463.1 | 2 | 314 | |||
| XM_011524162.2 | XP_011522464.1 | 2 | 314 | |||
| XM_011524163.2 | XP_011522465.1 | 2 | 314 | |||
| XM_011524164.2 | XP_011522466.1 | 2 | 310 | |||
| XM_011524165.2 | XP_011522467.1 | 2 | 315 | |||
| XM_011524166.2 | XP_011522468.1 | 2 | 313 | |||
| XM_011524167.2 | XP_011522469.1 | 2 | 312 | |||

ARL17A
Genomic Map