siRNA Details
| Gene Symbol: ECSCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077693.3 | NP_001071161.1 | 9 | 629 | ||
| NM_001293739.1 | NP_001280668.1 | 9 | 629 | |||
| NR_121659.1 | 10 | 738 | ||||
| GenBank mRNA | BC146912.1 | - | 9 | 601 | 9056840 | |
| BC146913.1 | - | 9 | 601 | 9056841 | ||
| BI834795.1 | - | 9 | 579 | |||
| DQ462572.1 | - | 9 | 623 | |||
| EU025066.1 | - | 9 | 629 | |||

ECSCR
Genomic Map