siRNA Details
Gene Symbol: LOC644794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136637.1 | 1 | 587 | |||
GenBank mRNA | AK301184.1 | - | 1 | 587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC644794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136637.1 | 1 | 587 | |||
GenBank mRNA | AK301184.1 | - | 1 | 587 |