siRNA Details
| Gene Symbol: FLJ16734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131514.1 | - | 1 | 1420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ16734| Gene Symbol: FLJ16734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131514.1 | - | 1 | 1420 | ||