siRNA Details
| Gene Symbol: LOC642249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724939.3 | XP_006725002.1 | 4 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC642249| Gene Symbol: LOC642249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724939.3 | XP_006725002.1 | 4 | 856 | ||