siRNA Details
| Gene Symbol: LOC101929231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110240.1 | 3 | 740 | |||
| GenBank mRNA | BG213919.1 | - | 1 | 211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929231| Gene Symbol: LOC101929231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110240.1 | 3 | 740 | |||
| GenBank mRNA | BG213919.1 | - | 1 | 211 | ||