siRNA Details
| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033829.1 | 5 | 492 | |||
| GenBank mRNA | AF339814.1 | - | 5 | 506 | ||
| BG215598.1 | - | 2 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR4500HG| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033829.1 | 5 | 492 | |||
| GenBank mRNA | AF339814.1 | - | 5 | 506 | ||
| BG215598.1 | - | 2 | 145 | |||