siRNA Details
| Gene Symbol: SPOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001664.2 | NP_001001664.1 | 11 | 1447 | ||
| XM_005263655.4 | XP_005263712.1 | 12 | 1410 | |||
| XM_005263656.4 | XP_005263713.1 | 12 | 1339 | |||
| XM_005263657.4 | XP_005263714.1 | 11 | 1252 | |||
| GenBank mRNA | BC071613.1 | - | 11 | 1426 | 5275316 | |
| BX538143.1 | - | 2 | 66 | |||

SPOPL
Genomic Map