siRNA Details
| Gene Symbol: TBC1D22A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122047.1 | 1 | 435 | |||
| GenBank mRNA | AK057318.1 | - | 1 | 413 | ||
| BC041833.1 | - | 1 | 435 | |||
| CR456602.1 | - | 1 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D22A-AS1| Gene Symbol: TBC1D22A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122047.1 | 1 | 435 | |||
| GenBank mRNA | AK057318.1 | - | 1 | 413 | ||
| BC041833.1 | - | 1 | 435 | |||
| CR456602.1 | - | 1 | 301 | |||