siRNA Details
| Gene Symbol: HOST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134505.1 | 1 | 313 | |||
| GenBank mRNA | BC033760.1 | - | 1 | 216 | ||
| Gene Symbol: UGT2B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265702.3 | XP_005265759.1 | 1 | 297 | ||

HOST2
Genomic Map