siRNA Details
Gene Symbol: LOC642980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131413.1 | - | 1 | 821 |
Gene Symbol: CXorf40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171909.3 | NP_001165380.1 | 5 | 3637 | ||
NM_001324276.1 | NP_001311205.1 | 6 | 3709 | |||
NM_001324279.1 | NP_001311208.1 | 6 | 3658 | |||
GenBank mRNA | AK131373.1 | - | 1 | 240 |
Gene Symbol: CXorf40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531181.2 | XP_011529483.1 | 6 | 3978 | ||
XM_017029590.1 | XP_016885079.1 | 6 | 4029 | |||
XM_017029591.1 | XP_016885080.1 | 6 | 4538 | |||
XM_017029592.1 | XP_016885081.1 | 6 | 3997 | |||
XM_017029593.1 | XP_016885082.1 | 7 | 4175 |
