siRNA Details
| Gene Symbol: LOC441155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271675.1 | NP_001258604.1 | 2 | 2255 | ||
| XM_017010889.1 | XP_016866378.1 | 2 | 1771 | |||
| GenBank mRNA | AK302514.1 | - | 2 | 2211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441155| Gene Symbol: LOC441155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271675.1 | NP_001258604.1 | 2 | 2255 | ||
| XM_017010889.1 | XP_016866378.1 | 2 | 1771 | |||
| GenBank mRNA | AK302514.1 | - | 2 | 2211 | ||