siRNA Details
| Gene Symbol: SMIM15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 496 | ||
| GenBank mRNA | AK291180.1 | - | 4 | 405 | ||
| BC014242.2 | - | 3 | 350 | |||
| BC047489.2 | - | 3 | 377 | |||
| BC066550.1 | - | 3 | 475 | |||
| BC103711.1 | - | 3 | 451 | |||
| BC146876.1 | - | 3 | 341 | 9056804 | ||
| BC146881.1 | - | 3 | 341 | 9056809 | ||
| BX538015.1 | - | 3 | 491 | |||
| BX648459.1 | - | 3 | 468 | |||
| DA039277.1 | - | 3 | 496 | |||

SMIM15
Genomic Map